siRNA Details
| Gene Symbol: POLG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001126131.1 | NP_001119603.1 | 10 | 2068 | ||
| NM_002693.2 | NP_002684.1 | 10 | 2080 | |||
| GenBank mRNA | AK291281.1 | - | 10 | 2099 | ||
| BC042571.1 | - | 10 | 2065 | 4384475 | ||
| BC050559.1 | - | 10 | 2049 | 6042595 | ||
| D84103.1 | - | 9 | 1904 | |||
| U60325.1 | - | 9 | 1957 | |||
| X98093.1 | - | 10 | 2080 | |||

POLG
Genomic Map