siRNA Details
| Gene Symbol: CAPN12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144691.4 | NP_653292.2 | 21 | 2697 | ||
| XM_017026355.1 | XP_016881844.1 | 19 | 2709 | |||
| XM_017026356.1 | XP_016881845.1 | 18 | 2646 | |||
| XM_017026357.1 | XP_016881846.1 | 20 | 2637 | |||
| XM_017026358.1 | XP_016881847.1 | 20 | 2634 | |||
| XM_017026360.1 | XP_016881849.1 | 16 | 2496 | |||
| XM_017026364.1 | XP_016881853.1 | 19 | 2345 | |||
| GenBank mRNA | AK125713.1 | - | 9 | 2108 | ||
| AK127398.1 | - | 21 | 2697 | |||

CAPN12
Genomic Map