siRNA Details
| Gene Symbol: CTSK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000396.3 | NP_000387.1 | 4 | 601 | ||
| GenBank mRNA | AK130134.1 | - | 4 | 420 | ||
| AK308229.1 | - | 3 | 262 | |||
| AK312880.1 | - | 4 | 418 | |||
| AU118043.1 | - | 4 | 497 | |||
| AU119985.1 | - | 4 | 496 | |||
| BC016058.1 | - | 4 | 483 | 4860926 | ||
| CR541675.1 | - | 3 | 372 | |||
| CR541719.1 | - | 3 | 372 | |||
| S79895.1 | - | 4 | 513 | |||
| U13665.1 | - | 4 | 477 | |||
| U20280.1 | - | 4 | 390 | |||

CTSK
Genomic Map