siRNA Details
| Gene Symbol: PEX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000318.2 | NP_000309.1 | 4 | 815 | ||
| NM_001079867.1 | NP_001073336.1 | 3 | 783 | |||
| NM_001172086.1 | NP_001165557.1 | 5 | 821 | |||
| NM_001172087.1 | NP_001165558.1 | 3 | 679 | |||
| GenBank mRNA | AK312997.1 | - | 2 | 482 | ||
| BC000661.2 | - | 4 | 588 | 3347824 | ||
| BC005375.1 | - | 3 | 584 | 3934225 | ||
| BC093043.1 | - | 4 | 601 | 30399827 | ||
| M85038.1 | - | 4 | 622 | |||
| M86852.1 | - | 4 | 730 | |||

PEX2
Genomic Map