siRNA Details
| Gene Symbol: IDS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000202.7 | NP_000193.1 | 9 | 1756 | ||
| NM_001166550.3 | NP_001160022.1 | 9 | 1712 | |||
| GenBank mRNA | AK294541.1 | - | 9 | 1667 | ||
| AK295815.1 | - | 2 | 399 | |||
| BX647357.1 | - | 9 | 1610 | |||
| M58342.1 | - | 9 | 1663 | |||

IDS
Genomic Map