siRNA Details
| Gene Symbol: IDS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006123.4 | NP_006114.1 | 8 | 1393 | ||
| NR_104128.1 | 9 | 1523 | ||||
| GenBank mRNA | BC006170.1 | - | 9 | 1381 | 4025980 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

IDS| Gene Symbol: IDS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006123.4 | NP_006114.1 | 8 | 1393 | ||
| NR_104128.1 | 9 | 1523 | ||||
| GenBank mRNA | BC006170.1 | - | 9 | 1381 | 4025980 | |