siRNA Details
| Gene Symbol: C9orf139 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207511.2 | NP_997394.1 | 2, 3 | 1675 | ||
| XM_017014717.1 | XP_016870206.1 | 2, 3 | 640 | |||
| GenBank mRNA | AK093587.1 | - | 1, 2 | 44 | ||
| AK124899.1 | - | 2, 3 | 1675 | |||
| BC132809.1 | - | 1, 2 | 449 | 40146831 | ||
| BC136839.1 | - | 1, 2 | 449 | 9020830 | ||

C9orf139
Genomic Map