siRNA Details
| Gene Symbol: FAM166A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001710.2 | NP_001001710.1 | 3 | 439 | ||
| XM_011518694.1 | XP_011516996.1 | 4 | 1424 | |||
| XM_011518696.2 | XP_011516998.1 | 4 | 1424 | |||
| XM_011518697.1 | XP_011516999.1 | 4 | 1424 | |||
| XM_017014718.1 | XP_016870207.1 | 3 | 1279 | |||
| GenBank mRNA | AK097419.1 | - | 3 | 417 | ||
| AY604177.1 | - | 3 | 417 | |||
| BC132916.1 | - | 3 | 384 | 40146938 | ||
| BC132918.1 | - | 3 | 384 | 40146940 | ||

FAM166A
Genomic Map