siRNA Details
| Gene Symbol: TPRN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001128228.2 | NP_001121700.2 | 4 | 2604 | ||
| GenBank mRNA | AK074735.1 | - | 3 | 2082 | ||
| BC023530.2 | - | 4 | 1612 | |||
| BC048807.1 | - | 4 | 1302 | |||
| BC071831.1 | - | 4 | 1934 | |||
| BC098411.1 | - | 4 | 2042 | |||
| BC111500.1 | - | 4 | 1733 | 4992533 | ||

TPRN
Genomic Map