siRNA Details
| Gene Symbol: UNC80 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032504.1 | NP_115893.1 | 36 | 5745 | ||
| NM_182587.3 | NP_872393.3 | 36 | 5730 | |||
| XM_005246476.1 | XP_005246533.1 | 37 | 5940 | |||
| XM_011511004.1 | XP_011509306.1 | 37 | 5985 | |||
| XM_011511005.1 | XP_011509307.1 | 37 | 5982 | |||
| XM_011511006.1 | XP_011509308.1 | 37 | 5979 | |||
| XM_011511007.1 | XP_011509309.1 | 37 | 5949 | |||
| XM_011511008.1 | XP_011509310.1 | 37 | 5985 | |||
| XM_011511010.2 | XP_011509312.1 | 37 | 5985 | |||
| XM_017003884.1 | XP_016859373.1 | 37 | 5970 | |||
| XM_017003885.1 | XP_016859374.1 | 37 | 5943 | |||
| XM_017003886.1 | XP_016859375.1 | 37 | 5985 | |||
| XM_017003887.1 | XP_016859376.1 | 36 | 5781 | |||
| XM_017003888.1 | XP_016859377.1 | 36 | 5739 | |||
| XM_017003889.1 | XP_016859378.1 | 36 | 5736 | |||
| XM_017003890.1 | XP_016859379.1 | 36 | 5724 | |||
| XM_017003891.1 | XP_016859380.1 | 36 | 6004 | |||
| XM_017003892.1 | XP_016859381.1 | 24 | 4788 | |||
| XM_017003893.1 | XP_016859382.1 | 37 | 5985 | |||
| XM_017003894.1 | XP_016859383.1 | 15 | 2216 | |||
| GenBank mRNA | AB058746.2 | - | 22 | 3190 | ||

UNC80
Genomic Map