siRNA Details
| Gene Symbol: KLHL40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152393.3 | NP_689606.2 | 1 | 1003 | ||
| GenBank mRNA | AK056577.1 | - | 1 | 985 | ||
| AK291294.1 | - | 1 | 985 | |||
| AK300682.1 | - | 2 | 309 | |||
| AY176040.1 | - | 1 | 1003 | |||
| AY177390.1 | - | 1 | 1003 | |||
| BC110491.2 | - | 1 | 971 | 40007554 | ||

KLHL40
Genomic Map