siRNA Details
| Gene Symbol: SLC25A53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012755.4 | NP_001012773.2 | 2 | 898 | ||
| XM_005262129.4 | XP_005262186.1 | 2 | 844 | |||
| XM_011530952.2 | XP_011529254.1 | 3 | 1041 | |||
| XM_011530953.2 | XP_011529255.1 | 3 | 872 | |||
| XM_017029514.1 | XP_016885003.1 | 3 | 1683 | |||
| XM_017029515.1 | XP_016885004.1 | 4 | 1191 | |||
| XM_017029516.1 | XP_016885005.1 | 3 | 1814 | |||
| GenBank mRNA | AL833609.1 | - | 2 | 899 | ||
| BC140812.1 | - | 1 | 758 | 9021680 | ||
| BC140814.1 | - | 1 | 758 | 9021682 | ||

SLC25A53
Genomic Map