siRNA Details
| Gene Symbol: STIM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277961.1 | NP_001264890.1 | 3 | 864 | ||
| NM_001277962.1 | NP_001264891.1 | 3 | 864 | |||
| NM_003156.3 | NP_003147.2 | 3 | 864 | |||
| GenBank mRNA | AK098480.1 | - | 2 | 344 | ||
| AK314928.1 | - | 3 | 385 | |||
| BC021300.2 | - | 3 | 856 | 4899542 | ||
| HQ317451.1 | - | 3 | 863 | |||
| JX014264.1 | - | 3 | 296 | |||
| U52426.1 | - | 3 | 861 | |||

STIM1
Genomic Map