siRNA Details
| Gene Symbol: RSPH4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001010892.2 | NP_001010892.1 | 3 | 1398 | ||
| NM_001161664.1 | NP_001155136.1 | 3 | 1398 | |||
| XM_017010826.1 | XP_016866315.1 | 4 | 4965 | |||
| GenBank mRNA | AK095293.1 | - | 4 | 1257 | ||
| AK299754.1 | - | 3 | 1398 | |||
| BC106949.2 | - | 3 | 1376 | 40034633 | ||

RSPH4A
Genomic Map