siRNA Details
| Gene Symbol: DUOXA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207581.3 | NP_997464.2 | 1 | 360 | ||
| XM_017022180.1 | XP_016877669.1 | 1 | 364 | |||
| GenBank mRNA | AK303955.1 | - | 1 | 359 | ||
| BC137465.1 | - | 1 | 281 | 9021465 | ||
| BX537581.1 | - | 1 | 211 | |||
| DB243076.1 | - | 1 | 359 | |||
| DC393748.1 | - | 1 | 360 | |||
| DQ489734.1 | - | 1 | 211 | |||

DUOXA2
Genomic Map