siRNA Details
| Gene Symbol: CCDC151 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001302453.1 | NP_001289382.1 | 8 | 916 | ||
| NM_001302454.1 | NP_001289383.1 | 6 | 1030 | |||
| NM_145045.4 | NP_659482.3 | 8 | 1210 | |||
| XM_017026241.1 | XP_016881730.1 | 8 | 1149 | |||
| GenBank mRNA | AK056363.1 | - | 8 | 1131 | ||
| AK294355.1 | - | 10 | 1110 | |||
| AK302113.1 | - | 8 | 917 | |||
| AK308072.1 | - | 6 | 992 | |||
| BC014252.2 | - | 7 | 767 | |||
| BC141828.1 | - | 8 | 1142 | 40148425 | ||
| BC142637.1 | - | 8 | 1155 | 40147788 | ||
| BC144421.1 | - | 6 | 689 | |||

CCDC151
Genomic Map