siRNA Details
| Gene Symbol: NHLRC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198586.2 | NP_940988.2 | 1 | 1066 | ||
| GenBank mRNA | AY324850.1 | - | 1 | 1066 | ||
| BC103888.1 | - | 1 | 1066 | 40004980 | ||
| BC103889.1 | - | 1 | 1066 | 40004983 | ||
| BC103890.1 | - | 1 | 1066 | 40004985 | ||

NHLRC1
Genomic Map