siRNA Details
| Gene Symbol: OR51A4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005329.1 | NP_001005329.1 | 1 | 635 | ||
| GenBank mRNA | BC140757.1 | - | 1 | 661 | 9021625 | |
| Gene Symbol: OR51A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004748.1 | NP_001004748.1 | 1 | 635 | ||

OR51A4
Genomic Map