siRNA Details
| Gene Symbol: LDLRAD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013693.2 | NP_001013715.2 | 1 | 254 | ||
| XM_005245873.4 | XP_005245930.1 | 1 | 310 | |||
| GenBank mRNA | AK127275.1 | - | 1 | 254 | ||
| BC146860.1 | - | 1 | 156 | 9056788 | ||

LDLRAD2
Genomic Map