siRNA Details
| Gene Symbol: CFAP99 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001193282.2 | NP_001180211.2 | 9 | 1012 | ||
| XM_011513472.2 | XP_011511774.1 | 9 | 1066 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CFAP99| Gene Symbol: CFAP99 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001193282.2 | NP_001180211.2 | 9 | 1012 | ||
| XM_011513472.2 | XP_011511774.1 | 9 | 1066 | |||