siRNA Details
| Gene Symbol: UNC93B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030930.3 | NP_112192.2 | 10 | 1463 | ||
| XM_011545290.1 | XP_011543592.1 | 8 | 1073 | |||
| XM_011545291.2 | XP_011543593.1 | 9 | 1355 | |||
| GenBank mRNA | AJ271326.1 | - | 10 | 1422 | ||
| AJ422142.1 | - | 10 | 1431 | |||
| AY007125.1 | - | 9 | 1250 | |||
| BC025669.1 | - | 10 | 1409 | |||
| BC033623.1 | - | 10 | 1461 | |||
| BC092472.1 | - | 10 | 1431 | 30348121 | ||
| BC101568.1 | - | 10 | 1407 | 8069074 | ||
| BC105104.1 | - | 10 | 1407 | 8144107 | ||

UNC93B1
Genomic Map