siRNA Details
| Gene Symbol: UFSP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001015072.3 | NP_001015072.2 | 1 | 704 | ||
| GenBank mRNA | AK292467.1 | - | 1 | 704 | ||
| BC017456.2 | - | 1 | 639 | |||
| BC067904.1 | - | 1 | 667 | 5310615 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

UFSP1| Gene Symbol: UFSP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001015072.3 | NP_001015072.2 | 1 | 704 | ||
| GenBank mRNA | AK292467.1 | - | 1 | 704 | ||
| BC017456.2 | - | 1 | 639 | |||
| BC067904.1 | - | 1 | 667 | 5310615 | ||