siRNA Details
| Gene Symbol: ZBTB34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099270.1 | NP_001092740.1 | 2 | 1370 | ||
| XM_005251989.4 | XP_005252046.1 | 3 | 1483 | |||
| XM_011518699.2 | XP_011517001.1 | 2 | 1386 | |||
| GenBank mRNA | AB082524.1 | - | 2 | 1370 | ||
| DQ227306.1 | - | 1 | 1285 | |||

ZBTB34
Genomic Map