siRNA Details
| Gene Symbol: CUEDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271875.1 | NP_001258804.1 | 2 | 605 | ||
| NM_001292025.1 | NP_001278954.1 | 2 | 910 | |||
| XM_006721900.2 | XP_006721963.1 | 2 | 1220 | |||
| XM_017024640.1 | XP_016880129.1 | 2 | 575 | |||
| XM_017024641.1 | XP_016880130.1 | 2 | 573 | |||
| GenBank mRNA | AK000746.1 | - | 2 | 910 | ||
| AK000977.1 | - | 1 | 344 | |||
| BC056882.1 | - | 2 | 605 | 6150503 | ||

CUEDC1
Genomic Map