siRNA Details
| Gene Symbol: C6orf201 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001085401.2 | NP_001078870.1 | 3, 4 | 1048 | ||
| NR_104464.1 | 2, 3 | 670 | ||||
| XM_011514613.2 | XP_011512915.1 | 3, 4 | 662 | |||
| XM_017010861.1 | XP_016866350.1 | 2, 3 | 679 | |||
| GenBank mRNA | AF429972.1 | - | 2, 3 | 498 | ||
| AK301893.1 | - | 2, 3 | 483 | |||
| BC047663.1 | - | 3, 4 | 1045 | 5266746 | ||
| BC066986.1 | - | 2, 3 | 659 | 4827732 | ||

C6orf201
Genomic Map