siRNA Details
Gene Symbol: C9orf106 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012715.3 | NP_001012733.2 | 2 | 411 | ||
GenBank mRNA | AK092588.1 | - | 2 | 412 | ||
BC132699.1 | - | 2 | 408 | 40146721 | ||
BC132701.1 | - | 2 | 408 | 40146723 |
