siRNA Details
| Gene Symbol: KRTAP5-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001480.2 | NP_001001480.2 | 1 | 958 | ||
| GenBank mRNA | AJ628247.1 | - | 3 | 910 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

KRTAP5-5| Gene Symbol: KRTAP5-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001001480.2 | NP_001001480.2 | 1 | 958 | ||
| GenBank mRNA | AJ628247.1 | - | 3 | 910 | ||