siRNA Details
| Gene Symbol: PRKCQ-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_036502.1 | 3 | 1142 | |||
| NR_036503.1 | 2 | 1078 | ||||
| GenBank mRNA | AY007155.1 | - | 2 | 964 | ||
| DB554966.1 | - | 2 | 388 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

PRKCQ-AS1| Gene Symbol: PRKCQ-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_036502.1 | 3 | 1142 | |||
| NR_036503.1 | 2 | 1078 | ||||
| GenBank mRNA | AY007155.1 | - | 2 | 964 | ||
| DB554966.1 | - | 2 | 388 | |||