siRNA Details
| Gene Symbol: IFIT1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001010987.2 | NP_001010987.1 | 2 | 1081 | ||
| GenBank mRNA | BC137368.1 | - | 2 | 1051 | 9021366 | |
| BC137369.1 | - | 2 | 1051 | 9021367 | ||
| BC150189.1 | - | 2 | 1063 | 40148511 | ||
| BX647300.1 | - | 2 | 1081 | |||

IFIT1B
Genomic Map