siRNA Details
| Gene Symbol: TMEM41B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015012.3 | NP_055827.1 | 7 | 1078 | ||
| NR_028491.2 | 7 | 1078 | ||||
| GenBank mRNA | AK303339.1 | - | 7 | 769 | ||
| BC035034.1 | - | 7 | 956 | 5259179 | ||
| BC044597.1 | - | 7 | 947 | 4811565 | ||
| BC064366.1 | - | 7 | 835 | |||
| BC091524.1 | - | 7 | 877 | 30512350 | ||
| D26067.1 | - | 7 | 884 | |||

TMEM41B
Genomic Map