siRNA Details
| Gene Symbol: TMEM41B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015012.3 | NP_055827.1 | 5 | 847 | ||
| NR_028491.2 | 5 | 847 | ||||
| GenBank mRNA | AK303339.1 | - | 5 | 538 | ||
| BC035034.1 | - | 5 | 725 | 5259179 | ||
| BC044597.1 | - | 5 | 716 | 4811565 | ||
| BC064366.1 | - | 5 | 604 | |||
| BC091524.1 | - | 5 | 646 | 30512350 | ||
| D26067.1 | - | 5 | 653 | |||
| DR157551.1 | - | 5 | 700 | |||

TMEM41B
Genomic Map