siRNA Details
| Gene Symbol: DBX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004329.2 | NP_001004329.2 | 3 | 711 | ||
| GenBank mRNA | AK131230.1 | - | 5 | 711 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

DBX2| Gene Symbol: DBX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004329.2 | NP_001004329.2 | 3 | 711 | ||
| GenBank mRNA | AK131230.1 | - | 5 | 711 | ||