siRNA Details
| Gene Symbol: LINC00238 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024338.2 | 2 | 227 | |||
| NR_110308.1 | 2 | 227 | ||||
| GenBank mRNA | BC033546.1 | - | 2 | 198 | ||
| BC056671.1 | - | 2 | 231 | |||
| BC070365.1 | - | 2 | 207 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00238| Gene Symbol: LINC00238 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_024338.2 | 2 | 227 | |||
| NR_110308.1 | 2 | 227 | ||||
| GenBank mRNA | BC033546.1 | - | 2 | 198 | ||
| BC056671.1 | - | 2 | 231 | |||
| BC070365.1 | - | 2 | 207 | |||