siRNA Details
| Gene Symbol: TTLL13P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104604.1 | 2 | 426 | |||
| GenBank mRNA | AK127965.1 | - | 2 | 402 | ||
| BC144453.1 | - | 2 | 176 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TTLL13P| Gene Symbol: TTLL13P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104604.1 | 2 | 426 | |||
| GenBank mRNA | AK127965.1 | - | 2 | 402 | ||
| BC144453.1 | - | 2 | 176 | |||