siRNA Details
| Gene Symbol: LOC440313 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK127131.1 | - | 1 | 2485 | ||
| BC140899.1 | - | 1 | 1528 | 9021767 | ||
| BC140900.1 | - | 1 | 1528 | 9021768 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC440313| Gene Symbol: LOC440313 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK127131.1 | - | 1 | 2485 | ||
| BC140899.1 | - | 1 | 1528 | 9021767 | ||
| BC140900.1 | - | 1 | 1528 | 9021768 | ||