siRNA Details
| Gene Symbol: SHISA9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145204.2 | NP_001138676.2 | 1 | 899 | ||
| NM_001145205.1 | NP_001138677.2 | 1 | 899 | |||
| XM_005255539.3 | XP_005255596.2 | 1 | 900 | |||
| XM_011522642.2 | XP_011520944.1 | 1 | 900 | |||
| GenBank mRNA | AK299610.1 | - | 2 | 329 | ||
| AK309038.1 | - | 1 | 240 | |||
| CR627300.1 | - | 1 | 95 | |||
| CR749545.1 | - | 1 | 95 | |||
| DC375222.1 | - | 2 | 330 | |||

SHISA9
Genomic Map