siRNA Details
| Gene Symbol: CTRB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025200.3 | NP_001020371.3 | 7 | 878 | ||
| GenBank mRNA | AK225933.1 | - | 7 | 865 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CTRB2| Gene Symbol: CTRB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025200.3 | NP_001020371.3 | 7 | 878 | ||
| GenBank mRNA | AK225933.1 | - | 7 | 865 | ||