siRNA Details
| Gene Symbol: MAFTRR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104663.1 | 6 | 530 | |||
| GenBank mRNA | BX108785.1 | - | 6 | 530 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MAFTRR| Gene Symbol: MAFTRR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104663.1 | 6 | 530 | |||
| GenBank mRNA | BX108785.1 | - | 6 | 530 | ||