siRNA Details
| Gene Symbol: ZNF506 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099269.2 | NP_001092739.1 | 4 | 716 | ||
| NM_001145404.1 | NP_001138876.1 | 3 | 620 | |||
| GenBank mRNA | AB209500.1 | - | 1 | 3722 | ||
| AK095575.1 | - | 4 | 716 | |||
| AK122848.1 | - | 3 | 620 | |||
| AL136548.1 | - | 4 | 674 | |||

ZNF506
Genomic Map