siRNA Details
| Gene Symbol: ZNF506 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099269.2 | NP_001092739.1 | 3 | 295 | ||
| GenBank mRNA | AK095575.1 | - | 3 | 295 | ||
| AL136548.1 | - | 3 | 253 | |||
| BC018100.1 | - | 3 | 267 | |||
| DA122515.1 | - | 3 | 295 | |||

ZNF506
Genomic Map