siRNA Details
| Gene Symbol: MINOS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001032363.3 | NP_001027535.1 | 4 | 1160 | ||
| NM_001204082.1 | NP_001191011.1 | 3 | 1112 | |||
| NM_001204083.1 | NP_001191012.1 | 5 | 1401 | |||
| NR_033757.2 | 5 | 1345 | ||||
| NR_033758.2 | 5 | 1239 | ||||
| GenBank mRNA | AK094318.1 | - | 4 | 1137 | ||

MINOS1
Genomic Map