siRNA Details
| Gene Symbol: LINC01191 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_034128.1 | 2 | 783 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01191| Gene Symbol: LINC01191 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_034128.1 | 2 | 783 | |||