siRNA Details
| Gene Symbol: CFAP99 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001193282.2 | NP_001180211.2 | 2 | 211 | ||
| XM_011513472.2 | XP_011511774.1 | 2 | 265 | |||
| GenBank mRNA | AK126028.1 | - | 2 | 182 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CFAP99| Gene Symbol: CFAP99 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001193282.2 | NP_001180211.2 | 2 | 211 | ||
| XM_011513472.2 | XP_011511774.1 | 2 | 265 | |||
| GenBank mRNA | AK126028.1 | - | 2 | 182 | ||