siRNA Details
| Gene Symbol: LOC441052 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134656.1 | 1 | 751 | |||
| GenBank mRNA | AF131741.1 | - | 1 | 751 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC441052| Gene Symbol: LOC441052 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134656.1 | 1 | 751 | |||
| GenBank mRNA | AF131741.1 | - | 1 | 751 | ||