siRNA Details
| Gene Symbol: MARCH11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001102562.1 | NP_001096032.1 | 3 | 953 | ||
| GenBank mRNA | BC150513.1 | - | 1 | 82 | ||
| BC150514.1 | - | 1 | 83 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MARCH11| Gene Symbol: MARCH11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001102562.1 | NP_001096032.1 | 3 | 953 | ||
| GenBank mRNA | BC150513.1 | - | 1 | 82 | ||
| BC150514.1 | - | 1 | 83 | |||