siRNA Details
Gene Symbol: C5orf56 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK096941.1 | - | 4 | 448 | ||
BC130299.1 | - | 3 | 346 | 40146330 | ||
BC130656.1 | - | 3 | 346 | 40146687 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: C5orf56 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK096941.1 | - | 4 | 448 | ||
BC130299.1 | - | 3 | 346 | 40146330 | ||
BC130656.1 | - | 3 | 346 | 40146687 |