siRNA Details
| Gene Symbol: LOC100289470 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK093197.1 | - | 2 | 1506 | ||
| Gene Symbol: C5orf60 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK093042.1 | - | 1 | 257 | ||

LOC100289470
Genomic Map