siRNA Details
| Gene Symbol: GTF2IP20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_132119.1 | 11 | 2900 | |||
| Gene Symbol: LOC107986805 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_017012932.1 | XP_016868421.1 | 1 | 2325 | ||

GTF2IP20
Genomic Map