siRNA Details
| Gene Symbol: LOC101927768 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125845.1 | 9 | 1678 | |||
| GenBank mRNA | BC042098.1 | - | 9 | 1678 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101927768| Gene Symbol: LOC101927768 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125845.1 | 9 | 1678 | |||
| GenBank mRNA | BC042098.1 | - | 9 | 1678 | ||