siRNA Details
| Gene Symbol: ZNF716 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159279.1 | NP_001152751.1 | 4 | 581 | ||
| GenBank mRNA | AK131575.1 | - | 4 | 581 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF716| Gene Symbol: ZNF716 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159279.1 | NP_001152751.1 | 4 | 581 | ||
| GenBank mRNA | AK131575.1 | - | 4 | 581 | ||